Få mere at vide
5-Methylcotinine-d3, TRC
Artikelnummer. 30468222
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30468222 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468222 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29452710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methylcotinine-d3
CAS 1217003-12-7
Molekylær formel C11H11D3N2O
Mængde 10 mg
Synonym 5-Methylcotinine D3
InChI formel InChI=1S/C11H14N2O/c1-8-5-9(7-12-6-8)10-3-4-11(14)13(10)2/h5-7,10H,3-4H2,1-2H3/t10-/m1/s1/i2D3
SMIL [2H]C([2H])([2H])N1[C@H](CCC1=O)c2cncc(C)c2
IUPAC navn (5R)-5-(5-methylpyridin-3-yl)-1-(trideuteriomethyl)pyrrolidin-2-one
Molekylvægt (g/mol) 193.26
Formel vægt 193.1294