Få mere at vide
5-Methyl Zolpidem, TRC
Artikelnummer. 17963725
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963725 10 mg
17911165 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17963725 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33860010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346600-85-8
Molekylær formel C19 H21 N3 O
Molekylvægt (g/mol) 307.39
Synonym N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide, Zolpidem Tartrate Imp. A (EP)
IUPAC navn N,N-dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
SMIL CN(C)C(=O)Cc1c(nc2cc(C)ccn12)c3ccc(C)cc3
Tekniske data
Kemisk navn eller materiale N,N-Dimethyl-2-[7-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetamide
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C19H21N3O/c1-13-5-7-15(8-6-13)19-16(12-18(23)21(3)4)22-10-9-14(2)11-17(22)20-19/h5-11H,12H2,1-4H3
Formel vægt 307.1685
Procent renhed >95%