Learn More
5-Methyl Tetrazole, TRC
Product Code. 30479987
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30479987 10 g 10g
1 options
Product Code. 30479987 Supplier Toronto Research Chemicals Supplier No. TRCM33025510G
 Restricted Item
Forventet forsendelse: 07-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methyl Tetrazole
CAS 4076-36-2
Molecular Formula C2H4N4
Quantity 10 g
Synonym 5-Methyl-1,2,3,4-tetrazole, 5-Methyl-1H-tetrazole, 5-Methyl-2H-tetrazole, 5-Methyltetrazole, NSC 11136, T 2588F
InChI Formula InChI=1S/C2H4N4/c1-2-3-5-6-4-2/h1H3,(H,3,4,5,6)
SMILES Cc1nnn[nH]1
IUPAC Name 5-methyl-1H-tetrazole
Molecular Weight (g/mol) 84.08
Formula Weight 84.0436