Få mere at vide
5-Methyl-d3-uridine-6-d1, CDN
Artikelnummer. 30326570
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326570 0.05 g -
30319681 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326570 Leverandør CDN Leverandørnr. CDND60560.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methyluridine uridine-d4
CAS 82845-85-0
Molekylær formel C10 2H4 H10 N2 O6
Mængde 0.05 g
Synonym Uridine-6-d, 5-(methyl-d3)- (9 CI), 5-Methyl-d3-uridine-6-d1, 5-Methyl uridine-d4, 5-Methyluridine-d4, 5-Methyluridine-D4 (uridine-D4)
InChI formel InChI=1 S/C10H14N2O6/c1-4-2-12(10(17)11-8(4)16)9-7(15)6(14)5(3-13)18-9/h2,5-7,9,13-15 H,3H2,1H3,(H,11,16,17)/t5-,6-,7-,9-/m1/s1/i1D3,2 D
SMIL [2 H]C1=C(C(=O)NC(=O)N1[C@@H]2 O[C@H](CO)[C@@H](O)[C@H]2 O)C([2 H])([2 H])[2 H]
IUPAC navn 6-deuterio-1-[(2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trideuteriomethyl)pyrimidine-2,4-dione
Molekylvægt (g/mol) 262.25
Formel vægt 262.1103 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity