Få mere at vide
5-Methyl-5-propyl-2-dioxanone-d3, TRC
Artikelnummer. 30467269
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30467269 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467269 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320577100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methyl-d3-5-propyl-2-dioxanone
CAS 1184973-36-1
Molekylær formel C8 D3 H11 O3
Mængde 100 mg
Synonym 5-Propyl-5-(trideuteriomethyl)-1,3-dioxan-2-one, 5-(Methyl-d3)-5-propyl-1,3-dioxan-2-one, 2-(Methyl-d3)-2-propyl-1,3-propanediol cyclic carbonate, Carbonic acid 2-(methyl-d3)-2-propyltrimethylene ester, 5-Methyl-5-propyl-1,3-dioxan-2-one-d3, 5-Methyl-5-propyl-1,3-dioxan-2-one-D3 (5-methyl-D3)
InChI formel InChI=1S/C8H14O3/c1-3-4-8(2)5-10-7(9)11-6-8/h3-6H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])C1(CCC)COC(=O)OC1
IUPAC navn 5-propyl-5-(trideuteriomethyl)-1,3-dioxan-2-one
Molekylvægt (g/mol) 161.21
Formel vægt 161.1131