Få mere at vide
5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride, TRC
Artikelnummer. 30466265
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466265 1 g
30483622 25 g
30474800 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30466265 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3300851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 720720-96-7
Molekylær formel C8H11ClN2O2S
Molekylvægt (g/mol) 234.7
Synonym 4,5,6,7-Tetrahydro-5-methylthiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride, 4,5,6,7-Tetrahydro-5-methylthiazolo[5,4-c]pyridine-2-carboxylic Acid Monochloride, 5-Methyl-4,5,6,7-tetrahydro[1,3]thiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
IUPAC navn 5-methyl-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-2-carboxylic acid;hydrochloride
SMIL O=C(O)C1=NC2=C(S1)CN(C)CC2.Cl
Tekniske data
Kemisk navn eller materiale 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
Mængde 1 g
InChI formel InChI=1S/C8H10N2O2S.ClH/c1-10-3-2-5-6(4-10)13-7(9-5)8(11)12;/h2-4H2,1H3,(H,11,12);1H
Formel vægt 234.023