missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-3-phenylisoxazole-4-carbonyl chloride, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Density | 1.2700g/mL |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 221.64 |
Percent Purity | 90% |
Chemical Name or Material | 5-Methyl-3-phenylisoxazole-4-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'