Få mere at vide
5-Methyl-3-heptyl Alcohol-d18 (mixture of stereoisomers), CDN
Artikelnummer. 30328441
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328441 0.25 g -
30318828 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328441 Leverandør CDN Leverandørnr. CDND76150.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methyl-3-heptanol-D18
CAS 2708283-13-8
Molekylær formel C8 D18 O
Mængde 0.25 g
Synonym 5-Methyl-3-heptanol-D18, 5-Methyl-3-heptyl alcohol-d18, 5-(Methyl-d3)-3-heptan-1,1,1,2,2,3,4,4,5,6,6,7,7,7-d14-ol-d
InChI formel InChI=1 S/C8H18O/c1-4-7(3)6-8(9)5-2/h7-9 H,4-6H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7 D,8 D,9 D
SMIL [2 H]OC([2 H])(C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,2,3,4,4,5,6,6,7,7,7-tetradecadeuterio-3-deuteriooxy-5-(trideuteriomethyl)heptane
Molekylvægt (g/mol) 148.3388
Formel vægt 148.2487 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity