Learn More
5-Methyl-3-heptanone, Sigma-Aldrich™
Product Code. 15840541
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity Unit Size
15840541 1 L 1 L
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15840541 Supplier Sigma Aldrich Supplier No. 8186631000
 Restricted Item
Forventet forsendelse: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 541-85-5
Melting Point -57°C
Density 0.82 g/cm3 at 20°C
Boiling Point 157°C to 162°C/1013 hPa
Molecular Formula CH3CH2COCH2CH(CH3)CH2CH3
MDL Number MFCD00009340
Quantity 1 L
Synonym 5-Methyl-3-heptanone, Ethyl amyl ketone, Ethyl isoamyl ketone, Isoamyl ethyl ketone, 5-Methyl-3-heptanone
Molecular Weight (g/mol) 128.21
Grade Research