missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole, 97%, Thermo Scientific™

Catalog Number p-7036166
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
10350-70-6
C8H7N3O
161.16
MFCD00464110
JZIKSOZOBQKFEX-UHFFFAOYSA-N
2771672
CC1=NC(=NO1)C1=CC=NC=C1
This item is not returnable. View return policy
10350-70-6
C8H7N3O
161.16
MFCD00464110
JZIKSOZOBQKFEX-UHFFFAOYSA-N
2771672
CC1=NC(=NO1)C1=CC=NC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 4-5-methyl-1,2,4-oxadiazol-3-yl pyridine, 5-methyl-3-4-pyridyl-1,2,4-oxadiazole, pyridine,4-5-methyl-1,2,4-oxadiazol-3-yl, pyridine, 4-5-methyl-1,2,4-oxadiazol-3-yl, 5-methyl-3-pyridin-4-yl-1,2,4-oxadiazole
IUPAC Name 5-methyl-3-pyridin-4-yl-1,2,4-oxadiazole
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-(4-pyridyl)-1,2,4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'