Få mere at vide
5-Methyl-2’-deoxy Cytidine-d3, TRC
Artikelnummer. 30468128
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30468128 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468128 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2959021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(Methyl-d3)-2'-deoxy Cytidine
CAS 1160707-78-7
Renhedsgrad noter HPLC
Molekylær formel C10H12D3N3O4
Mængde 1 mg
Synonym Cytidine, 2'-deoxy-5-(methyl-d3)-, 2'-Deoxy-5-(methyl-d3)cytidine, 2'-Deoxy-5-methylcytidine-d3, 5-Methyl-2'-deoxycytidine-D3 (5-methyl-D3), 5-Methyldeoxycytidine-d3
InChI formel InChI=1S/C10H15N3O4/c1-5-3-13(10(16)12-9(5)11)8-2-6(15)7(4-14)17-8/h3,6-8,14-15H,2,4H2,1H3,(H2,11,12,16)/t6-,7+,8+/m0/s1/i1D3
SMIL [2H]C([2H])([2H])C1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N=C1N
Molekylvægt (g/mol) 244.26
Procent renhed >95