Learn More
Alfa Aesar™ 5-Methyl-2-(2-thienyl)benzimidazole, 95%
Brand: Alfa Aesar™ H64938.MD
Additional Details : CAS Number : 3878-24-8 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
3878-24-8 | |
214.286 | |
QIYVOSKFSQXPCG-UHFFFAOYSA-N | |
924806 | |
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3 |
C12H10N2S | |
MFCD06312144 | |
6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole | |
6-methyl-2-thiophen-2-yl-1H-benzimidazole |
Specifications
5-Methyl-2-(2-thienyl)benzimidazole | |
3878-24-8 | |
MFCD06312144 | |
QIYVOSKFSQXPCG-UHFFFAOYSA-N | |
6-methyl-2-thiophen-2-yl-1H-benzimidazole | |
924806 | |
Solid |
250mg | |
C12H10N2S | |
6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole | |
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3 | |
214.286 | |
214.29 | |
95% |