Få mere at vide
(5-Methyl-1H-imidazol-4-yl)methanol Hydrochloride, Mikromol™
Artikelnummer. 17970786
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17970786 25 mg
1 options
Artikelnummer. 17970786 Leverandør Mikromol Leverandørnr. MM0020.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(5-Methyl-1H-imidazol-4-yl)methanol Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (5-Methyl-1H-imidazol-4-yl)methanol Hydrochloride
CAS 38585-62-5
Analyt- eller komponentnavne (5-Methyl-1H-imidazol-4-yl)methanol Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C5 H8 N2 O . Cl H
Certificeringer/compliance ISO 17025
Synonym 1H-Imidazole-5-methanol, 4-methyl-, hydrochloride (1:1), 1H-Imidazole-4-methanol, 5-methyl-, monohydrochloride (9CI), (4-Methyl-1H-imidazol-5-yl)methanol hydrochloride, (5-Methyl-1H-imidazol-4-yl)methanol hydrochloride, 4-(Hydroxymethyl)-5-methylimidazole hydrochloride, 4-Methyl-1H-imidazole-5-methanol hydrochloride, 4-Methyl-5-imidazolemethanol hydrochloride, 5-Methyl-4-imidazolemethanol hydrochloride, Cimetidine Impurity I as Hydrochloride
InChI formel InChI=1S/C5H8N2O.ClH/c1-4-5(2-8)7-3-6-4;/h3,8H,2H2,1H3,(H,6,7);1H
SMIL Cl.Cc1[nH]cnc1CO
IUPAC navn (5-methyl-1H-imidazol-4-yl)methanol;hydrochloride
Molekylvægt (g/mol) 148.59
Mængde 25 mg
Formel vægt 148.0403 g/mol