Få mere at vide
5-methyl-1-(2-methylpropyl)-1H-pyrazol-4-amine, TRC
Artikelnummer. 30466766
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466766 10 mg
30482277 100 mg
30481355 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30466766 Leverandør Toronto Research Chemicals Leverandørnr. TRCM35604010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 405548-39-2
Molekylær formel C8 H15 N3
Molekylvægt (g/mol) 153.23
Synonym 1-Isobutyl-5-methyl-1H-pyrazol-4-amine, 4-Amino-1-isobutyl-5-methylpyrazole
IUPAC navn 1-isobutyl-5-methyl-pyrazol-4-amine
SMIL CC(C)Cn1ncc(N)c1C
Tekniske data
Kemisk navn eller materiale 5-methyl-1-(2-methylpropyl)-1H-pyrazol-4-amine
Mængde 10 mg
InChI formel InChI=1S/C8H15N3/c1-6(2)5-11-7(3)8(9)4-10-11/h4,6H,5,9H2,1-3H3
Formel vægt 153.1266