Få mere at vide
5-Methoxyuracil, TRC
Artikelnummer. 30471516
Skift visning
Mængde:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471516 1 g
30464447 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471516 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2717501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6623-81-0
Molekylær formel C5 H6 N2 O3
Molekylvægt (g/mol) 142.11
Synonym 5-Methoxy-2,4(1H,3H)-pyrimidinedione, Uracil, 5-methoxy- (6CI,7CI,8CI), 5-Methoxypyrimidine-2,4-diol, 5-Methoxyuracil, NSC 55452
IUPAC navn 5-methoxy-1H-pyrimidine-2,4-dione
SMIL COC1=CNC(=O)NC1=O
Tekniske data
Kemisk navn eller materiale 5-Methoxyuracil
Mængde 1 g
InChI formel InChI=1S/C5H6N2O3/c1-10-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)
Formel vægt 142.0378