missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Methoxytryptamine, TRC

Artikelnummer. 30472776
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30472776 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30472776 Leverandør Toronto Research Chemicals Leverandørnr. TRCM271695100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 5-Methoxytryptamine
CAS 608-07-1
Renhedsgrad noter HPLC
Molekylær formel C11 H14 N2 O
Mængde 100 mg
Synonym 2-(5-Methoxy-1H-indol-3-yl)ethanamine, 5-Methoxytryptamine, Melatonin Related Compound A (USP), 1H-Indole-3-ethanamine','5-methoxy- (9CI','ACI), 5-Methoxy-1H-indole-3-ethanamine (ACI), Indole','3-(2-aminoethyl)-5-methoxy- (6CI','8CI), 2-(5-Methoxy-1H-indol-3-yl)ethan-1-amine, 2-(5-Methoxyindol-3-yl)ethylamine, 2-[5-(Methyloxy)-1H-indol-3-yl]ethanamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-MeOT, 5-MOT, [2-(5-Methoxy-1H-indol-3-yl)ethyl]amine, Deacetylmelatonin, MeSH ID: D008735, Methoxytryptamine, NSC 56422
InChI formel InChI=1S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4-5,12H2,1H3
SMIL COc1ccc2[nH]cc(CCN)c2c1
IUPAC navn 2-(5-methoxy-1H-indol-3-yl)ethanamine
Molekylvægt (g/mol) 190.24
Formel vægt 190.1106
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.