Få mere at vide
5-Methoxytryptamine-Alpha,Alpha,Beta,Beta-d4, TRC
Artikelnummer. 30476474
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30476474 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476474 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2716961MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methoxytryptamine-alpha,alpha,beta,beta-d4
CAS 96236-05-4
Molekylær formel C11 D4 H10 N2 O
Mængde 1 mg
Synonym 1H-Indole-3-ethan-α,α,β,β-d4-amine','5-methoxy- (9CI), 5-Methoxytryptamine-α,α,β,β-D4, 5-Methoxytryptamine-α,α,β,β-d4, 5-Methoxytryptamine-d4, 5-Methoxy-1H-indole-3-ethan-α,α,β,β-d4-amine
InChI formel InChI=1S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4-5,12H2,1H3/i4D2,5D2
SMIL [2H]C([2H])(N)C([2H])([2H])c1c[nH]c2ccc(OC)cc12
IUPAC navn 1,1,2,2-tetradeuterio-2-(5-methoxy-1H-indol-3-yl)ethanamine
Molekylvægt (g/mol) 194.266
Formel vægt 194.136