Få mere at vide
5-Methoxypyrimidine-2,4(1H,3H)-dione (5-Methoxyuracil), Mikromol™
Artikelnummer. 30052556
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052556 100 mg
30050086 25 mg
30458277 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30052556 Leverandør Mikromol Leverandørnr. MM0593.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Methoxypyrimidine-2,4(1H,3H)-dione (5-Methoxyuracil)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methoxyuracil
CAS 6623-81-0
Analyt- eller komponentnavne 5-Methoxyuracil
Forsendelsestilstand Room Temperature
Molekylær formel C5 H6 N2 O3
Certificeringer/compliance ISO 9001
Synonym 5-Methoxy-2,4(1H,3H)-pyrimidinedione, Uracil, 5-methoxy- (6CI,7CI,8CI), 5-Methoxypyrimidine-2,4-diol, 5-Methoxyuracil, NSC 55452, Fluorouracil Imp. D (EP), Fluorouracil Impurity D
InChI formel InChI=1S/C5H6N2O3/c1-10-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)
SMIL COC1=CNC(=O)NC1=O
IUPAC navn 5-methoxy-1H-pyrimidine-2,4-dione
Molekylvægt (g/mol) 142.11
Mængde 100 mg
Formel vægt 142.0378 g/mol