missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Methoxycarbonyl Methyl Uridine, TRC

Artikelnummer. 30467828
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467828 5 mg
30481981 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30467828 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2616155MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 29428-50-0
Molekylær formel C12H16N2O8
Molekylvægt (g/mol) 316.26
Synonym 1,2,3,4-Tetrahydro-2,4-dioxo-1-β-D-ribofuranosyl-5-pyrimidineacetic Acid Methyl Ester, 5-Carboxymethyluridine Methyl Ester
SMIL O[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N(C=C2CC(OC)=O)C(NC2=O)=O

Tekniske data

Kemisk navn eller materiale 5-Methoxycarbonyl Methyl Uridine
Mængde 5 mg
InChI formel InChI=1S/C12H16N2O8/c1-21-7(16)2-5-3-14(12(20)13-10(5)19)11-9(18)8(17)6(4-15)22-11/h3,6,8-9,11,15,17-18H,2,4H2,1H3,(H,13,19,20)/t6-,8-,9-,11-/m1/s1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.