missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxybenzofuroxan, 97%, Thermo Scientific™

Catalog Number p-7021475
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7791-49-3
C7H6N2O3
166.136
MFCD00068052
ABSKHUSUHODMEL-UHFFFAOYSA-N
5-methoxybenzofurazan-1-oxide, 5-methoxy-2,1,3-benzoxadiazole 1-oxide, 5-methoxy-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methoxybenzofuroxane, 5-methoxybenzofurazane 1-oxide, 5-methoxybenzofurazan, 1-oxide, 5-methoxybenzo c 1,2,5-oxadiazol-1-ol, 2,1,3-benzoxadiazole,5-methoxy-,1-oxide, 2,1,3-benzoxadiazole, 5-methoxy-, monooxide
321028
5-methoxy-1-oxido-2,1,3-benzoxadiazol-1-ium
COC1=CC2=NO[N+](=C2C=C1)[O-]
This item is not returnable. View return policy
7791-49-3
C7H6N2O3
166.136
MFCD00068052
ABSKHUSUHODMEL-UHFFFAOYSA-N
5-methoxybenzofurazan-1-oxide, 5-methoxy-2,1,3-benzoxadiazole 1-oxide, 5-methoxy-2,1,3-benzoxadiazol-1-ium-1-olate, 5-methoxybenzofuroxane, 5-methoxybenzofurazane 1-oxide, 5-methoxybenzofurazan, 1-oxide, 5-methoxybenzo c 1,2,5-oxadiazol-1-ol, 2,1,3-benzoxadiazole,5-methoxy-,1-oxide, 2,1,3-benzoxadiazole, 5-methoxy-, monooxide
321028
5-methoxy-1-oxido-2,1,3-benzoxadiazol-1-ium
COC1=CC2=NO[N+](=C2C=C1)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 112°C to 114°C
Quantity 5 g
Beilstein 643053
Formula Weight 166.14
Percent Purity 97%
Chemical Name or Material 5-Methoxybenzofuroxan
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'