Få mere at vide
5-(Methoxy-d3)-2-mercaptobenzimidazole, TRC
Artikelnummer. 30473186
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30473186 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473186 Leverandør Toronto Research Chemicals Leverandørnr. TRCM262762100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-(Methoxy-d3)-2-mercaptobenzimidazole
CAS 922730-86-7
Molekylær formel C8H5D3N2OS
Mængde 100 mg
Synonym 1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione-d3, 2-Mercapto-5-methoxy-1H-benzimidazole-d3, 2-Mercapto-5-methoxybenzimidazole-d3, 5-Methoxy-1H-benzimidazole-2-thiol-d3, 5-Methoxy-2-benzimidazolethiol-d3
InChI formel InChI=1S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)10-8(12)9-6/h2-4H,1H3,(H2,9,10,12)/i1D3
SMIL SC1=NC2=C(N1)C=CC(OC([2H])([2H])[2H])=C2
IUPAC navn 5-(methoxy-d3)-1H-benzo[d]imidazole-2-thiol
Molekylvægt (g/mol) 183.25
Formel vægt 183.05