Få mere at vide
5-Methoxy-Alpha-oxo-6-(phenylmethoxy)-1H-indole-3-acetamide, TRC
Artikelnummer. 30475384
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475384 25 mg
30475688 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30475384 Leverandør Toronto Research Chemicals Leverandørnr. TRCM27154025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methoxy-alpha-oxo-6-(phenylmethoxy)-1H-indole-3-acetamide
CAS 4790-22-1
Molekylær formel C18 H16 N2 O4
Mængde 25 mg
Synonym 6-(Benzyloxy)-5-methoxyindole-3-glyoxylamide
InChI formel InChI=1S/C18H16N2O4/c1-23-15-7-12-13(17(21)18(19)22)9-20-14(12)8-16(15)24-10-11-5-3-2-4-6-11/h2-9,20H,10H2,1H3,(H2,19,22)
SMIL COc1cc2c(c[nH]c2cc1OCc3ccccc3)C(=O)C(=O)N
IUPAC navn 2-(5-methoxy-6-phenylmethoxy-1H-indol-3-yl)-2-oxoacetamide
Molekylvægt (g/mol) 324.331
Formel vægt 324.111