missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Isobutyl-4-nitro-1H-pyrazole-3-carboxylic Acid, TRC

Artikelnummer. 30406696
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30406696 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30406696 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7806251G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 5-Isobutyl-4-nitro-1H-pyrazole-3-carboxylic Acid
CAS 222729-55-7
Molekylær formel C8H11N3O4
Mængde 1 g
Synonym 5-(2-Methylpropyl)-4-nitro-1H-pyrazole-3-carboxylic Acid
InChI formel InChI=1S/C8H11N3O4/c1-4(2)3-5-7(11(14)15)6(8(12)13)10-9-5/h4H,3H2,1-2H3,(H,9,10)(H,12,13)
SMIL CC(C)Cc1[nH]nc(C(=O)O)c1[N+](=O)[O-]
IUPAC navn 5-(2-methylpropyl)-4-nitro-1H-pyrazole-3-carboxylic acid
Molekylvægt (g/mol) 213.19
Formel vægt 213.075
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.