missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodouracil, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is used as an intermediate. It has an antitumor activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an intermediate. It has an antitumor activity.
Solubility
Very faint turbidity in NH3aq. Soluble in 1M NaOH.
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an intermediate. It has an antitumor activity.
Solubility
Very faint turbidity in NH3aq. Soluble in 1M NaOH.
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 274°C to 276°C |
Quantity | 10 g |
Beilstein | 4891 |
Sensitivity | Light sensitive |
Solubility Information | Very faint turbidity in NH3aq. Soluble in 1M NaOH. |
Formula Weight | 237.99 |
Percent Purity | 97% |
Chemical Name or Material | 5-Iodouracil |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'