missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodoisatin, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.
Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.
Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | ∼276°C (decomposition) |
Quantity | 5 g |
Beilstein | 131248 |
Sensitivity | Light sensitive |
Solubility Information | Soluble acetone,acetic acid,ethanol. Insoluble in water. |
Formula Weight | 273.03 |
Percent Purity | 97% |
Chemical Name or Material | 5-Iodoisatin |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'