Få mere at vide
5'-Hydroxyphenyl Carvedilol-d5, TRC
Artikelnummer. 30405798
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30405798 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405798 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94912610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5'-Hydroxyphenyl Carvedilol-d5
CAS 1189704-21-9
Molekylær formel C24H21D5N2O5
Mængde 10 mg
Synonym 3-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxypropyl-d5]amino]ethoxy]-4-methoxy-phenol, 5-Hydroxycarvedilol-d5, BM 140830-d5
InChI formel InChI=1S/C24H26N2O5/c1-29-21-10-9-16(27)13-23(21)30-12-11-25-14-17(28)15-31-22-8-4-7-20-24(22)18-5-2-3-6-19(18)26-20/h2-10,13,17,25-28H,11-12,14-15H2,1H3/i14D2,15D2,17D
SMIL OC1=CC(OCCNC([2H])([2H])C([2H])(O)C([2H])([2H])OC2=CC=CC3=C2C4=C(N3)C=CC=C4)=C(OC)C=C1
Molekylvægt (g/mol) 427.5
Formel vægt 427.2156