Få mere at vide
5-Hydroxypentanal, TRC
Artikelnummer. 30403773
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30403773 50 mg
30402481 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30403773 Leverandør Toronto Research Chemicals Leverandørnr. TRCH99993550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Hydroxypentanal
CAS 4221-03-8
Molekylær formel C5H10O2
Mængde 50 mg
InChI formel InChI=1S/C5H10O2/c6-4-2-1-3-5-7/h4,7H,1-3,5H2
SMIL OCCCCC=O
IUPAC navn 5-hydroxypentanal
Molekylvægt (g/mol) 102.13
Formel vægt 102.0681