Få mere at vide
5-(Hydroxymethyl)-2,4-dimethylpyridin-3-ol Hydrochloride, Mikromol™
Artikelnummer. 30460371
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460371 4 x 25 mg
17974276 100 mg
17991726 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460371 Leverandør Mikromol Leverandørnr. MM0021.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(Hydroxymethyl)-2,4-dimethylpyridin-3-ol Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Deoxypyridoxine hydrochloride
CAS 148-51-6
Analyt- eller komponentnavne 4-Deoxypyridoxine hydrochloride
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C8 H11 N O2 . Cl H
Certificeringer/compliance ISO 17025
Synonym 2,4-Dimethyl-3-hydroxy-5-hydroxymethylpyridine Hydrochloride, 4-Deoxypyridoxine Hydrochloride, 3-Hydroxy-5-hydroxymethyl-2,4-dimethylpyridine Hydrochloride, 4-Deoxypyridoxol Hydrochloride, 5-Hydroxy-4,6-dimethyl-3-pyridinemethanol Hydrochloride, 4-Desoxyadermin Hydrochloride, 4-Desoxypyridoxine Hydrochloride, 5-Hydroxymethyl-2,4-dimethylpyridin-3-ol Hydrochloride, Pyridoxine Hydrochloride Imp. B (EP), Pyridoxine Imp. B (EP), 5-(Hydroxymethyl)-2,4-dimethylpyridin-3-ol Hydrochloride, Pyridoxine Hydrochloride Imp. B (EP) as Hydrochloride, Pyridoxine Hydrochloride Impurity B as Hydrochloride, Pyridoxine Impurity B as Hydrochloride
InChI formel InChI=1S/C8H11NO2.ClH/c1-5-7(4-10)3-9-6(2)8(5)11;/h3,10-11H,4H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.Cc1ncc(CO)c(C)c1O
IUPAC navn 5-(hydroxymethyl)-2,4-dimethylpyridin-3-ol;hydrochloride
Molekylvægt (g/mol) 189.64
Mængde 4 x 25 mg
Formel vægt 189.0557 g/mol