Få mere at vide
5-Hydroxyindole-3-acetic Acid-D2, TRC
Artikelnummer. 30404117
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30404117 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404117 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9536081MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Hydroxyindole-3-acetic acid 2,2-d2
CAS 56209-31-5
Molekylær formel C10 2H2 H7 N O3
Mængde 1 mg
Synonym 1H-Indole-3-acetic-α,α-d2 acid, 5-hydroxy- (9CI), 5-Hydroxy-1H-indole-3-acetic-α,α-d2 acid, 5-Hydroxyindole-3-acetic-2,2-d2 acid, 5-Hydroxyindole-3-acetic acid-d2, 5-Hydroxyindole-3-acetic-2,2-D2 acid, 5-HIAA-d2
InChI formel InChI=1S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12H,3H2,(H,13,14)/i3D2
SMIL [2H]C([2H])(C(=O)O)c1c[nH]c2ccc(O)cc12
IUPAC navn 2,2-dideuterio-2-(5-hydroxy-1H-indol-3-yl)acetic acid
Molekylvægt (g/mol) 193.2
Formel vægt 193.0708