Få mere at vide
5-Hydroxyindole-2,4,6,7-d4-3-acetic-2,2-d2 Acid, CDN
Artikelnummer. 30317381
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317381 0.005 g
30311938 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317381 Leverandør CDN Leverandørnr. CDND79550.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Hydroxyindole-2,4,6,7-d4-3-acetic-2,2-d2 acid
CAS 2748469-82-9
Molekylær formel C10 D6 H3 N O3
Mængde 0.005 g
Synonym 5-Hydroxyindole-3-acetic acid-D6, 5-Hydroxyindole-2,4,6,7-D4-3-acetic-D2 acid, 5-Hydroxy-1 H-indole-2,4,6,7-d4-3-acetic-α,α-d2 acid, 5-Hydroxy-2,4,6,7-d4-indole-3-acetic-2,2-d2 acid, 5-HIAA-d6
InChI formel InChI=1 S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12 H,3H2,(H,13,14)/i1D,2 D,3D2,4 D,5 D
SMIL [2 H]c1[nH]c2c([2 H])c([2 H])c(O)c([2 H])c2c1C([2 H])([2 H])C(=O)O
IUPAC navn 2,2-dideuterio-2-(2,4,6,7-tetradeuterio-5-hydroxy-1 H-indol-3-yl)acetic acid
Molekylvægt (g/mol) 197.2203
Formel vægt 197.0959 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity