missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Hydroxyhexanoic Acid Sodium Salt, TRC

Artikelnummer. 30391919
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30391919 2.5 g
30390602 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30391919 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9428252.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 185956-02-9
Molekylær formel C6H11NaO3
Molekylvægt (g/mol) 154.14
Synonym 5-Hydroxycaproic Acid
IUPAC navn sodium 5-hydroxyhexanoate
SMIL OC(CCCC(O[Na])=O)C

Tekniske data

Kemisk navn eller materiale 5-Hydroxyhexanoic Acid Sodium Salt
Mængde 2.5 g
InChI formel InChI=1S/C6H12O3.Na/c1-5(7)3-2-4-6(8)9;/h5,7H,2-4H2,1H3,(H,8,9);/q;+1/p-1
Formel vægt 154.06
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.