Få mere at vide
5-Hydroxy Tryptophol-d4, TRC
Artikelnummer. 30403419
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30403419 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403419 Leverandør Toronto Research Chemicals Leverandørnr. TRCH97600250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Hydroxy Tryptophol-d4
CAS 66640-87-7
Molekylær formel C10 2H4 H7 N O2
Mængde 50 mg
Synonym 1H-Indole-3-ethan-α,α,β,β-d4-ol, 5-hydroxy- (9CI), 5-Hydroxy-1H-indole-3-ethanol-D4 (ethanol-D4), 5-Hydroxy tryptophol-d4, 5-Hydroxytryptophol-d4, 2-(5-Hydroxyindol-3-yl)ethanol-1,1,2,2-d4, 5-Hydroxy-3-(2-hydroxy-1,1,2,2-tetradeuteroethyl)indole
InChI formel InChI=1S/C10H11NO2/c12-4-3-7-6-11-10-2-1-8(13)5-9(7)10/h1-2,5-6,11-13H,3-4H2/i3D2,4D2
SMIL [2H]C([2H])(O)C([2H])([2H])c1c[nH]c2ccc(O)cc12
IUPAC navn 3-(1,1,2,2-tetradeuterio-2-hydroxyethyl)-1H-indol-5-ol
Molekylvægt (g/mol) 181.22
Formel vægt 181.1041