Få mere at vide
5-Hydroxy Thiabendazole-13C2,15N, TRC
Artikelnummer. 30134972
Skift visning
Mængde:
0.25 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134972 5 mg
30003255 0.25 mg
30009305 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134972 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9612025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391052-65-5
Molekylær formel C813C2H7N215NOS
Molekylvægt (g/mol) 220.23
Synonym 2-(4-Thiazolyl)-1H-benzimidazol-6-ol-13C2,15N, 5-Hydroxythiabendazole-13C2,15N;
SMIL OC1=CC2=C(N=[13C]([13C]3=CSC=N3)[15NH]2)C=C1
Tekniske data
Kemisk navn eller materiale 5-Hydroxy Thiabendazole-13C2,15N
Mængde 5 mg
InChI formel InChI=1S/C10H7N3OS/c14-6-1-2-7-8(3-6)13-10(12-7)9-4-15-5-11-9/h1-5,14H,(H,12,13)/i9+1,10+1,13+1
Formel vægt 220.0347