missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-p-naphthoquinone, 97%, Thermo Scientific Chemicals

Catalog Number p-146649
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
481-39-0
C10H6O3
174.16
MFCD00001684
KQPYUDDGWXQXHS-UHFFFAOYSA-N
juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
3806
CHEBI:15794
5-hydroxynaphthalene-1,4-dione
OC1=CC=CC2=C1C(=O)C=CC2=O
This item is not returnable. View return policy
481-39-0
C10H6O3
174.16
MFCD00001684
KQPYUDDGWXQXHS-UHFFFAOYSA-N
juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
3806
CHEBI:15794
5-hydroxynaphthalene-1,4-dione
OC1=CC=CC2=C1C(=O)C=CC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 161.0°C to 163.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 08, 308
Merck Index 15, 5317
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in aq.sol. of alkalies,soluble in alcohol and ether
Formula Weight 174.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-p-naphthoquinone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'