Få mere at vide
5-Hydroxy Flunixin-d3 See H942424, TRC
Artikelnummer. 30406905
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30406905 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406905 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9424221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Hydroxy Flunixin-d3
CAS 1185088-54-3
Molekylær formel C14H8D3F3N2O3
Mængde 1 mg
Synonym 5-Hydroxy-2-[[2-methyl-d3-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic Acid
InChI formel InChI=1S/C14H11F3N2O3/c1-7-10(14(15,16)17)3-2-4-11(7)19-12-9(13(21)22)5-8(20)6-18-12/h2-6,20H,1H3,(H,18,19)(H,21,22)/i1D3
SMIL OC1=CN=C(NC2=C(C([2H])([2H])[2H])C(C(F)(F)F)=CC=C2)C(C(O)=O)=C1
Molekylvægt (g/mol) 315.26
Formel vægt 315.091