Få mere at vide
5-Hydroxy Buspirone-d8, TRC
Artikelnummer. 30405047
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405047 1 mg
30404830 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405047 Leverandør Toronto Research Chemicals Leverandørnr. TRCH8307971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1330164-16-3
Molekylær formel C21H23D8N5O3
Molekylvægt (g/mol) 409.55
Synonym 8-[4-[4-(5-Hydroxy-2-pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione-d8
SMIL O=C(CC1(CCCC1)CC2=O)N2C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N3CCN(C4=NC=C(O)C=N4)CC3
Tekniske data
Kemisk navn eller materiale 5-Hydroxy Buspirone-d8
Mængde 1 mg
InChI formel InChI=1S/C21H31N5O3/c27-17-15-22-20(23-16-17)25-11-9-24(10-12-25)7-3-4-8-26-18(28)13-21(14-19(26)29)5-1-2-6-21/h15-16,27H,1-14H2/i3D2,4D2,7D2,8D2
Formel vægt 409.2929