Få mere at vide
5-Hydroxy-2-methyl-1H-imidazole-1-ethanol, TRC
Artikelnummer. 30406709
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30406709 50 mg
30405972 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30406709 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94750550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 138169-63-8
Molekylær formel C6 H10 N2 O2
Molekylvægt (g/mol) 142.16
IUPAC navn 3-(2-hydroxyethyl)-2-methyl-imidazol-4-ol
SMIL Cc1ncc(O)n1CCO
Tekniske data
Kemisk navn eller materiale 5-Hydroxy-2-methyl-1H-imidazole-1-ethanol
Mængde 50 mg
InChI formel InChI=1S/C6H10N2O2/c1-5-7-4-6(10)8(5)2-3-9/h4,9-10H,2-3H2,1H3
Formel vægt 142.0742