Få mere at vide
5-Hydroxy-2-[1-methyl-1-[[benzylcarbamoyl]amino]ethyl]-6-methoxypyrimidine-4-carboxylic Acid Methyl Ester-d3, TRC
Artikelnummer. 30403640
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30403640 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403640 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9482421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Hydroxy-2-(1-methyl-1-((benzylcarbamoyl)amino)ethyl)-6-methoxypyrimidine-4-carboxylic Acid Methyl Ester-d3
CAS 1185032-53-4
Molekylær formel C18 H18 D3 N3 O6
Mængde 1 mg
Synonym 5-Hydroxy-6-(methoxy-d3)-2-[1-methyl-1-[[(phenylmethoxy)carbonyl]amino]ethyl]-4-pyrimidinecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C18H21N3O6/c1-18(2,21-17(24)27-10-11-8-6-5-7-9-11)16-19-12(15(23)26-4)13(22)14(20-16)25-3/h5-9,22H,10H2,1-4H3,(H,21,24)/i3D3
SMIL [2H]C([2H])([2H])Oc1nc(nc(C(=O)OC)c1O)C(C)(C)NC(=O)OCc2ccccc2
IUPAC navn methyl 5-hydroxy-2-[2-(phenylmethoxycarbonylamino)propan-2-yl]-6-(trideuteriomethoxy)pyrimidine-4-carboxylate
Molekylvægt (g/mol) 378.39
Formel vægt 378.1619