Få mere at vide
5-Hydroxy-1-methyl-4-nitroimidazole Sodium Salt (>90%), TRC
Artikelnummer. 30137145
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137145 500 mg
17902915 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137145 Leverandør Toronto Research Chemicals Leverandørnr. TRCH947755500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35681-68-6
Molekylær formel C4H4N3NaO3
Molekylvægt (g/mol) 165.08
Synonym 1H-Imidazol-5-ol, 1-methyl-4-nitro-, sodium salt (1:1) (ACI), 1H-Imidazol-5-ol, 1-methyl-4-nitro-, sodium salt (9CI), 1-Methyl-4-nitro-1H-imidazol-5-ol sodium salt, 1-Methyl-4-nitro-5-hydroxyimidazole sodium salt, 5-Hydroxy-1-methyl-4-nitroimidazole sodium salt, 1-Methyl-4-nitro-1H-imidazol-5-ol Sodium Salt, Azathioprine BP Impurity E
IUPAC navn sodium;3-methyl-5-nitroimidazol-4-olate
SMIL [Na+].Cn1cnc(c1[O-])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 5-Hydroxy-1-methyl-4-nitroimidazole Sodium Salt
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C4H5N3O3.Na/c1-6-2-5-3(4(6)8)7(9)10;/h2,8H,1H3;/q;+1/p-1
Formel vægt 165.015
Procent renhed >95%