missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenyl acetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7035719
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5048-26-0
C8H14O2
142.20
MFCD00060856
MPLWNENKBSBMFN-UHFFFAOYSA-N
5-hexenyl acetate, hex-5-en-1-yl acetate, acetic acid 5-hexenyl ester, 5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, acetic acid 5-hexen-1-yl ester, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, 5-hexenyl acetate #, acmc-1aw0j
537536
hex-5-enyl acetate
CC(=O)OCCCCC=C
This item is not returnable. View return policy
5048-26-0
C8H14O2
142.20
MFCD00060856
MPLWNENKBSBMFN-UHFFFAOYSA-N
5-hexenyl acetate, hex-5-en-1-yl acetate, acetic acid 5-hexenyl ester, 5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, acetic acid 5-hexen-1-yl ester, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, 5-hexenyl acetate #, acmc-1aw0j
537536
hex-5-enyl acetate
CC(=O)OCCCCC=C

(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.

Solubility
Insoluble in water, soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.883
Boiling Point 173°C to 174°C
Flash Point 60°C (142°F)
Linear Formula CH3CO2(CH2)4CH=CH2
Refractive Index 1.423
Quantity 5 g
UN Number UN3272
Solubility Information Insoluble in water,soluble in alcohol.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 5-Hexenyl acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'