missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexenyl acetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.
Solubility
Insoluble in water, soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.
Solubility
Insoluble in water, soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Density | 0.883 |
Boiling Point | 173°C to 174°C |
Flash Point | 60°C (142°F) |
Linear Formula | CH3CO2(CH2)4CH=CH2 |
Refractive Index | 1.423 |
Quantity | 100 g |
UN Number | UN3272 |
Solubility Information | Insoluble in water,soluble in alcohol. |
Formula Weight | 142.2 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'