missing translation for 'onlineSavingsMsg'
Learn More

5-Hex-1-ynylnicotinic acid, 97%, Thermo Scientific™

Catalog Number p-2702846
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
306935-31-9
C12H13NO2
203.241
MFCD00830967
FKWDNFDBDZUPIQ-UHFFFAOYSA-N
5-hex-1-ynylnicotinic acid, 5-hex-1-ynylnicotinc acid, 3-pyridinecarboxylicacid, 5-1-hexyn-1-yl, 5-hex-1-yn-1-yl pyridine-3-carboxylic acid, 5-hex-1-ynyl-nicotinic acid, 5-hex-1-ynyl nicotinic acid, 5-hex-1-yn-1-yl nicotinic acid
2736516
5-hex-1-ynylpyridine-3-carboxylic acid
CCCCC#CC1=CC(=CN=C1)C(=O)O
This item is not returnable. View return policy
306935-31-9
C12H13NO2
203.241
MFCD00830967
FKWDNFDBDZUPIQ-UHFFFAOYSA-N
5-hex-1-ynylnicotinic acid, 5-hex-1-ynylnicotinc acid, 3-pyridinecarboxylicacid, 5-1-hexyn-1-yl, 5-hex-1-yn-1-yl pyridine-3-carboxylic acid, 5-hex-1-ynyl-nicotinic acid, 5-hex-1-ynyl nicotinic acid, 5-hex-1-yn-1-yl nicotinic acid
2736516
5-hex-1-ynylpyridine-3-carboxylic acid
CCCCC#CC1=CC(=CN=C1)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 5-Hex-1-ynylnicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'