Få mere at vide
5-Formyltetrahydropteroic Acid, TRC
Artikelnummer. 17967694
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17967694 2.5 mg 2.5mg
17918964 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17967694 Leverandør Toronto Research Chemicals Leverandørnr. TRCF7010802.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4349-43-3
Molekylær formel C15 H16 N6 O4
Molekylvægt (g/mol) 344.33
Synonym Benzoic acid, 4-[[(2-amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]-, Benzoic acid, 4-[[(2-amino-5-formyl-1,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]- (9CI), Benzoic acid, p-[[(2-amino-5-formyl-5,6,7,8-tetrahydro-4-hydroxy-6-pteridinyl)methyl]amino]- (6CI,7CI,8CI), 5-Formyl-5,6,7,8-tetrahydropteroic acid, 5-Formyltetrahydropteroic acid
IUPAC navn 4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoic acid
SMIL NC1=NC(=O)C2=C(NCC(CNc3ccc(cc3)C(=O)O)N2C=O)N1
Tekniske data
Kemisk navn eller materiale 5-Formyltetrahydropteroic Acid
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C15H16N6O4/c16-15-19-12-11(13(23)20-15)21(7-22)10(6-18-12)5-17-9-3-1-8(2-4-9)14(24)25/h1-4,7,10,17H,5-6H2,(H,24,25)(H4,16,18,19,20,23)
Formel vægt 344.1233
Procent renhed >95%