Få mere at vide
5-Formylcytidine, TRC
Artikelnummer. 30136562
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136562 5 mg
30135735 10 mg
30010727 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136562 Leverandør Toronto Research Chemicals Leverandørnr. TRCF6989655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148608-53-1
Molekylær formel C10H13N3O6
Molekylvægt (g/mol) 271.23
Synonym 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidine-5-carbaldehyde;
SMIL O[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C([H])=O)C(N)=NC2=O
Tekniske data
Kemisk navn eller materiale 5-Formylcytidine
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C10H13N3O6/c11-8-4(2-14)1-13(10(18)12-8)9-7(17)6(16)5(3-15)19-9/h1-2,5-7,9,15-17H,3H2,(H2,11,12,18)/t5-,6-,7-,9-/m1/s1
Procent renhed >95%