Learn More
5-Fluorouracil-6-d1, TRC
Product Code. 30397983
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30397983 5 mg 5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397983 Supplier Toronto Research Chemicals Supplier No. TRCF5960035MG
 Restricted Item
Forventet forsendelse: 03-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Fluorouracil-6-d1
CAS 90344-84-6
Molecular Formula C4H2DFN2O2
Quantity 5 mg
Synonym 2,4(1H,3H)-Pyrimidinedione-6-d, 5-fluoro- (9CI), 5-Fluorouracil-6-d, 5-Fluoro-2,4(1H,3H)-pyrimidinedione-6-d, 2,4-Dihydroxy-5-fluoropyrimidine-6-d, Fluorouracil-6-D
SMILES [2H]C1=C(F)C(=O)NC(=O)N1
Molecular Weight (g/mol) 131.08
Formula Weight 131.0241