Få mere at vide
5-Fluorouracil-15N2, TRC
Artikelnummer. 30399623
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30399623 10 mg
30406457 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30399623 Leverandør Toronto Research Chemicals Leverandørnr. TRCF59600610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68941-95-7
Molekylær formel C4H3F(15N)2O2
Molekylvægt (g/mol) 132.06
Synonym 5-Fluoropyrimidine-2,4(1H,3H)-dione-1,3-15N2
IUPAC navn 5-fluoropyrimidine-2,4(1H,3H)-dione-1,3-15N2
SMIL O=C1C(F)=C[15NH]C(=O)[15NH]1
Tekniske data
Kemisk navn eller materiale 5-Fluorouracil-15N2
Mængde 10 mg
InChI formel InChI=1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)/i6+1,7+1
Formel vægt 132.01