Få mere at vide
5-Fluorodihydropyrimidine-2,4-dione, TRC
Artikelnummer. 30004642
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004642 10 mg
30009919 25 mg
30007907 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004642 Leverandør Toronto Research Chemicals Leverandørnr. TRCF59102510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 696-06-0
Molekylær formel C4 H5 F N2 O2
Molekylvægt (g/mol) 132.093
Synonym 5,6-Dihydro-5-fluorouracil, 2,4(1H,3H)-Pyrimidinedione, 5-fluorodihydro-, 5-Fluorodihydro-2,4(1H,3H)-pyrimidinedione (ACI), Hydrouracil, 5-fluoro- (6CI, 7CI, 8CI), 5,6-Dihydro-5-fluorouracil, 5-Fluoro-5,6-dihydrouracil, 5-Fluorodihydrouracil, 5-Fluorohydrouracil, Dihydro-5-fluorouracil
IUPAC navn 5-fluoro-1,3-diazinane-2,4-dione
SMIL FC1CNC(=O)NC1=O
Tekniske data
Kemisk navn eller materiale 5-Fluorodihydropyrimidine-2,4-dione
Mængde 10 mg
InChI formel InChI=1S/C4H5FN2O2/c5-2-1-6-4(9)7-3(2)8/h2H,1H2,(H2,6,7,8,9)
Formel vægt 132.034