missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Catalog Number p-7031207
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1092460-88-2
C8H5BrF4O
273.025
MFCD11519337
VUUGICQPCOWAIC-UHFFFAOYSA-N
2-trifluoromethoxy-5-fluorobenzyl bromide, 5-fluoro-2-trifluoromethoxy benzyl bromide, 2-bromomethyl-4-fluoro-1-trifluoromethoxy benzene
53421164
2-(bromomethyl)-4-fluoro-1-(trifluoromethoxy)benzene
C1=CC(=C(C=C1F)CBr)OC(F)(F)F
This item is not returnable. View return policy
1092460-88-2
C8H5BrF4O
273.025
MFCD11519337
VUUGICQPCOWAIC-UHFFFAOYSA-N
2-trifluoromethoxy-5-fluorobenzyl bromide, 5-fluoro-2-trifluoromethoxy benzyl bromide, 2-bromomethyl-4-fluoro-1-trifluoromethoxy benzene
53421164
2-(bromomethyl)-4-fluoro-1-(trifluoromethoxy)benzene
C1=CC(=C(C=C1F)CBr)OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Fluoro-2-(trifluoromethoxy)benzyl bromide, is used as a primary and secondary intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Refractive Index 1.466
Quantity 250 mg
UN Number UN3265
Formula Weight 273.02
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethoxy)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'