missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7024331
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
393-09-9
C7H3F4NO2
209.1
MFCD00039741
WMQOSURXFLBTPC-UHFFFAOYSA-N
5-fluoro-2-nitrobenzotrifluoride, 4-fluoro-1-nitro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl nitrobenzene, 2-nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 2-nitro-5-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl-4-nitrobenzene, 2-trifluoromethyl-4-fluoronitrobenzene, benzene, 4-fluoro-1-nitro-2-trifluoromethyl, 4-fluoro-1-nitro-2-trifluoromethyl-benzene, 2-nitro-5-fluorotrifluorotoluol
223094
4-fluoro-1-nitro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1F)C(F)(F)F)[N+](=O)[O-]
This item is not returnable. View return policy
393-09-9
C7H3F4NO2
209.1
MFCD00039741
WMQOSURXFLBTPC-UHFFFAOYSA-N
5-fluoro-2-nitrobenzotrifluoride, 4-fluoro-1-nitro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl nitrobenzene, 2-nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 2-nitro-5-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl-4-nitrobenzene, 2-trifluoromethyl-4-fluoronitrobenzene, benzene, 4-fluoro-1-nitro-2-trifluoromethyl, 4-fluoro-1-nitro-2-trifluoromethyl-benzene, 2-nitro-5-fluorotrifluorotoluol
223094
4-fluoro-1-nitro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1F)C(F)(F)F)[N+](=O)[O-]

5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 22°C to 24°C
Density 1.497
Boiling Point 87°C to 88°C (2 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.46
Quantity 5 g
UN Number UN2811
Beilstein 3304470
Solubility Information Soluble in chloroform and methanol.
Formula Weight 209.1
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-nitrobenzotrifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'