missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Fluoro-2-methoxybenzeneboronic acid pinacol ester, 96%
Brand: Alfa Aesar™ H62701.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1383806-53-8 | |
252.092 | |
2-5-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-fluoro-2-methoxyphenylboronic acid pinacol ester, amtb632, 5-fluoro-2-methoxybenzeneboronic acid pinacol ester, 2-2-methoxy-5-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
2-(5-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
C13H18BFO3 | |
HFSNSSDHOJSZKI-UHFFFAOYSA-N | |
69867754 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)OC |
Specifications
5-Fluoro-2-methoxybenzeneboronic acid pinacol ester | |
1383806-53-8 | |
2-5-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-fluoro-2-methoxyphenylboronic acid pinacol ester, amtb632, 5-fluoro-2-methoxybenzeneboronic acid pinacol ester, 2-2-methoxy-5-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC(=C2)F)OC | |
252.092 | |
252.09 | |
96% |
1g | |
C13H18BFO3 | |
HFSNSSDHOJSZKI-UHFFFAOYSA-N | |
2-(5-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
69867754 | |
Solid |